Anti-VEGFA Mouse Monoclonal Antibody [clone: VG-1]

Lieferant: Abcam
ABCAAB1316-100EA 519.88 CHF
ABCAAB1316-100
Anti-VEGFA Mouse Monoclonal Antibody [clone: VG-1]
Antikörper
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone: VG-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR