Anti-DDDDK tag Goat polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB1257-100EA 958 CHF
ABCAAB1257-100
Anti-DDDDK tag Goat polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: DDDDK tag (DDDDK tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR