Anti-Myc Tag Goat Polyclonal Antibody (Agarose)
Lieferant: Abcam
ABCAAB1253-100EA 524.65 CHF
ABCAAB1253-100
Anti-Myc Tag Goat Polyclonal Antibody (Agarose)
Antikörper
Type: Primary
Antigen: Myc Tag (Myc tag)
Clonality: polyclonal
Clone:
Conjugation: Agarose
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now