Anti-PPP2R1A Rabbit polyclonal antibody

ABCAAB1180-200EA 744 CHF
ABCAAB1180-200
Anti-PPP2R1A Rabbit polyclonal antibody
Type: Primary
Antigen: PPP2R1A (protein phosphatase 2, regulatory subunit A, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR