Anti-CD19 Mouse monoclonal antibody FITC (Fluorescein) [clone: CB19]

ABCAAB1167-500EA 294 CHF
ABCAAB1167-500
Anti-CD19 Mouse monoclonal antibody FITC (Fluorescein) [clone: CB19]
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone: CB19
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR