Anti-CD4 Mouse monoclonal antibody FITC (Fluorescein) [clone: EDU-2]

ABCAAB1154-500EA 321 CHF
ABCAAB1154-500
Anti-CD4 Mouse monoclonal antibody FITC (Fluorescein) [clone: EDU-2]
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone: EDU-2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR