Anti-MUC16 Mouse Monoclonal Antibody [clone: X75]

Supplier: Abcam
ABCAAB1107-100EA 933 CHF
ABCAAB1107-100
Anti-MUC16 Mouse Monoclonal Antibody [clone: X75]
Antibodies
Type: Primary
Antigen: MUC16 (mucin 16, cell surface associated)
Clonality: monoclonal
Clone: X75
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR