Anti-RAF1 Rabbit polyclonal antibody

ABCAAB1095-1EA 0 CHF
ABCAAB1095-1
Anti-RAF1 Rabbit polyclonal antibody
Type: Primary
Antigen: RAF1 (v-raf-1 murine leukemia viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR