Anti-Ketanserin Mouse monoclonal antibody

ABCAAB1061-250EA 691 CHF
ABCAAB1061-250
Anti-Ketanserin Mouse monoclonal antibody
Type: Primary
Antigen: Ketanserin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR