Anti-Hepatitis G Virus Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB1037-50EA 472.05 CHF
ABCAAB1037-50
Anti-Hepatitis G Virus Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Hepatitis G Virus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: unknown
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR