Anti-Hydroxyprogesterone Sheep polyclonal antibody

ABCAAB1027EA 660 CHF
ABCAAB1027
Anti-Hydroxyprogesterone Sheep polyclonal antibody
Type: Primary
Antigen: Hydroxyprogesterone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR