Anti-SNX1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB995-50EA 529.11 CHF
ABCAAB995-50
Anti-SNX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SNX1 (sorting nexin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR