Anti-CD68 Mouse Monoclonal Antibody [clone: KP1]

Supplier: Abcam
ABCAAB955-500EA 1000 CHF
ABCAAB955-500
Anti-CD68 Mouse Monoclonal Antibody [clone: KP1]
Antibodies
Type: Primary
Antigen: CD68 (CD68 molecule)
Clonality: monoclonal
Clone: KP1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR