Anti-S100 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB948-6EA 920 CHF
ABCAAB948-6
Anti-S100 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: S100
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR