Anti-MB Rabbit polyclonal antibody

ABCAAB943-6EA 708 CHF
ABCAAB943-6
Anti-MB Rabbit polyclonal antibody
Type: Primary
Antigen: MB (myoglobin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR