Anti-Lambda Light chain Rabbit polyclonal antibody

ABCAAB941-6EA 744 CHF
ABCAAB941-6
Anti-Lambda Light chain Rabbit polyclonal antibody
Type: Primary
Antigen: Lambda Light chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR