Anti-IgG Rabbit Polyclonal Antibody

ABCAAB938-6EA 737 CHF
ABCAAB938-6
Anti-IgG Rabbit Polyclonal Antibody
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR