Anti-Placental alkaline phosphatase (PLAP) Mouse monoclonal antibody [clone: 8B6]

ABCAAB913-6EA 708 CHF
ABCAAB913-6
Anti-Placental alkaline phosphatase (PLAP) Mouse monoclonal antibody [clone: 8B6]
Type: Primary
Antigen: Placental alkaline phosphatase (PLAP)
Clonality: monoclonal
Clone: 8B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR