Anti-KRT20 Mouse Monoclonal Antibody [clone: Ks20.8]

Supplier: Abcam
ABCAAB854-500EA 529.11 CHF
ABCAAB854-500
Anti-KRT20 Mouse Monoclonal Antibody [clone: Ks20.8]
Antibodies
Type: Primary
Antigen: KRT20 (keratin 20)
Clonality: monoclonal
Clone: Ks20.8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR