Anti-CD5 Mouse Monoclonal Antibody [clone: 4C7]

Supplier: Abcam
ABCAAB851-250EA 492.8 CHF
ABCAAB851-250
Anti-CD5 Mouse Monoclonal Antibody [clone: 4C7]
Antibodies
Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: monoclonal
Clone: 4C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR