Anti-UB Rabbit polyclonal antibody

ABCAAB837-500EA 708 CHF
ABCAAB837-500
Anti-UB Rabbit polyclonal antibody
Type: Primary
Antigen: UB (ubiquitin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR