Anti-RNA polymerase II CTD repeat YSPTSPS Mouse Monoclonal Antibody [clone: 8WG16]

Supplier: Abcam
ABCAAB817-100EA 975 CHF
ABCAAB817-100
Anti-RNA polymerase II CTD repeat YSPTSPS Mouse Monoclonal Antibody [clone: 8WG16]
Antibodies
Type: Primary
Antigen: RNA polymerase II CTD repeat YSPTSPS
Clonality: monoclonal
Clone: 8WG16
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR