Anti-MelanA Mouse Monoclonal Antibody [clone: A103]

Supplier: Abcam
ABCAAB785-500EA 518.74 CHF
ABCAAB785-500
Anti-MelanA Mouse Monoclonal Antibody [clone: A103]
Antibodies
Type: Primary
Antigen: MelanA
Clonality: monoclonal
Clone: A103
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR