Anti-Lambda Light chain Mouse Monoclonal Antibody [clone: N10/2]

Supplier: Abcam
ABCAAB780-500EA 903 CHF
ABCAAB780-500
Anti-Lambda Light chain Mouse Monoclonal Antibody [clone: N10/2]
Antibodies
Type: Primary
Antigen: Lambda Light chain
Clonality: monoclonal
Clone: N10/2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR