Anti-KRT8 Mouse Monoclonal Antibody [clone: 35bH11]
Lieferant: Abcam
ABCAAB758-500EA 524.65 CHF
ABCAAB758-500
Anti-KRT8 Mouse Monoclonal Antibody [clone: 35bH11]
Antikörper
Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: monoclonal
Clone: 35bH11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human