Anti-Cytokeratin Mouse monoclonal antibody unconjugated [clone: MNF116]

Supplier: Abcam
ABCAAB756-500EA 529.11 CHF
ABCAAB756-500
Anti-Cytokeratin Mouse monoclonal antibody unconjugated [clone: MNF116]
Antibodies
Type: Primary
Antigen: Cytokeratin
Clonality: monoclonal
Clone: MNF116
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR