Anti-CAPN2 Mouse Monoclonal Antibody [clone: 1E8]

Supplier: Abcam
ABCAAB713-100EA 911 CHF
ABCAAB713-100
Anti-CAPN2 Mouse Monoclonal Antibody [clone: 1E8]
Antibodies
Type: Primary
Antigen: CAPN2 (calpain 2, (m/II) large subunit)
Clonality: monoclonal
Clone: 100000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR