Anti-Prion protein PrP Mouse monoclonal antibody [clone: 1E7/D3]

ABCAAB706-50EA 672 CHF
ABCAAB706-50
Anti-Prion protein PrP Mouse monoclonal antibody [clone: 1E7/D3]
Type: Primary
Antigen: Prion protein PrP
Clonality: monoclonal
Clone: 1E7/D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR