Anti-Prion protein PrP Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB703-50EA 508.36 CHF
ABCAAB703-50
Anti-Prion protein PrP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Prion protein PrP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR