Anti-CCL16 Goat polyclonal antibody

ABCAAB626-100EA 691 CHF
ABCAAB626-100
Anti-CCL16 Goat polyclonal antibody
Type: Primary
Antigen: CCL16 (chemokine (C-C motif) ligand 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: unknown
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR