Anti-IgG Mouse Monoclonal Antibody [clone: hIgG1]

Lieferant: Abcam
ABCAAB436-250EA 214.63 CHF
ABCAAB436-250
Anti-IgG Mouse Monoclonal Antibody [clone: hIgG1]
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: hIgG1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR