Anti-IgE Mouse Monoclonal Antibody [clone: hIgE2]

Supplier: Abcam
ABCAAB435-250EA 207.49 CHF
ABCAAB435-250
Anti-IgE Mouse Monoclonal Antibody [clone: hIgE2]
Antibodies
Type: Secondary
Antigen: IgE
Clonality: Monoclonal
Clone: hIgE2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR