Anti-Isopeptide Mouse monoclonal antibody [clone: 71A3F1]

ABCAAB423-200EA 691 CHF
ABCAAB423-200
Anti-Isopeptide Mouse monoclonal antibody [clone: 71A3F1]
Type: Primary
Antigen: Isopeptide
Clonality: monoclonal
Clone: 71A3F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR