Anti-MB Mouse monoclonal antibody [clone: Myo2]

ABCAAB406-100EA 672 CHF
ABCAAB406-100
Anti-MB Mouse monoclonal antibody [clone: Myo2]
Type: Primary
Antigen: MB (myoglobin)
Clonality: monoclonal
Clone: Myo2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR