Anti-GFP Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB290-50EA 629.58 CHF
ABCAAB290-50
Anti-GFP Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR