Anti-ITGA4 Mouse Monoclonal Antibody [clone: 44H6]

Supplier: Abcam
ABCAAB220-100EA 570.61 CHF
ABCAAB220-100
Anti-ITGA4 Mouse Monoclonal Antibody [clone: 44H6]
Antibodies
Type: Primary
Antigen: ITGA4 (integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor))
Clonality: monoclonal
Clone: 44H6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR