Anti-MRE11A Mouse Monoclonal Antibody [clone: 12D7]

Supplier: Abcam
ABCAAB214-100EA 430.55 CHF
ABCAAB214-100
Anti-MRE11A Mouse Monoclonal Antibody [clone: 12D7]
Antibodies
Type: Primary
Antigen: MRE11A (MRE11 meiotic recombination 11 homolog A (S. cerevisiae))
Clonality: monoclonal
Clone: 12D7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR