Anti-TSG101 Mouse Monoclonal Antibody [clone: 4A10]
Lieferant: Abcam
ABCAAB83-100EA 581.89 CHF
ABCAAB83-100
Anti-TSG101 Mouse Monoclonal Antibody [clone: 4A10]
Antikörper
Type: Primary
Antigen: TSG101 (tumor susceptibility 101)
Clonality: monoclonal
Clone: 4A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat