Anti-CCNB1 Mouse monoclonal antibody unconjugated [clone: V152]

Supplier: Abcam
ABCAAB72-100EA 555.05 CHF
ABCAAB72-100
Anti-CCNB1 Mouse monoclonal antibody unconjugated [clone: V152]
Antibodies
Type: Primary
Antigen: CCNB1 (cyclin B1)
Clonality: monoclonal
Clone: V152
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR