Anti-ORC1 Mouse Monoclonal Antibody [clone: 7F6/1]

Supplier: Abcam
ABCAAB60-50EA 912 CHF
ABCAAB60-50
Anti-ORC1 Mouse Monoclonal Antibody [clone: 7F6/1]
Antibodies
Type: Primary
Antigen: ORC1 (origin recognition complex, subunit 1)
Clonality: monoclonal
Clone: 7F6/1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR