Anti-ERCC5 Mouse Monoclonal Antibody [clone: 8H7]

Supplier: Abcam
ABCAAB46-50EA 492.8 CHF
ABCAAB46-50
Anti-ERCC5 Mouse Monoclonal Antibody [clone: 8H7]
Antibodies
Type: Primary
Antigen: ERCC5 (excision repair cross-complementation group 5)
Clonality: monoclonal
Clone: 8H7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR