Anti-MYC Mouse Monoclonal Antibody [clone: 9E10]

Supplier: Abcam
ABCAAB32-100EA 591.36 CHF
ABCAAB32-100
Anti-MYC Mouse Monoclonal Antibody [clone: 9E10]
Antibodies
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone: 90000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR