Anti-EGFR Mouse monoclonal antibody unconjugated [clone: EGFR1]

Supplier: Abcam
ABCAAB30-100EA 544.67 CHF
ABCAAB30-100
Anti-EGFR Mouse monoclonal antibody unconjugated [clone: EGFR1]
Antibodies
Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: monoclonal
Clone: EGFR1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR