Anti-CD45RO Mouse Monoclonal Antibody [clone: UCH-L1]
Lieferant: Abcam
ABCAAB23-100EA 457.88 CHF
ABCAAB23-100
Anti-CD45RO Mouse Monoclonal Antibody [clone: UCH-L1]
Antikörper
Type: Primary
Antigen: CD45RO (CD45RO)
Clonality: monoclonal
Clone: UCH-L1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human