Anti-HDAC1 Rabbit polyclonal antibody unconjugated

Lieferant: Biorbyt
BIRBORB99450-20EA 0 CHF
BIRBORB99450-20 BIRBORB99450-100
Anti-HDAC1 Rabbit polyclonal antibody unconjugated
Antikörper
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: HDAC1 (histone deacetylase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR