Anti-GATA4 Rabbit polyclonal antibody unconjugated

Supplier: Biorbyt
BIRBORB99436-100EA 0 CHF
BIRBORB99436-100
Anti-GATA4 Rabbit polyclonal antibody unconjugated
Antibodies
Western Blot: 1:500-1:2000; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: GATA4 (GATA binding protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR