Anti-RAF1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB99406-20EA 0 CHF
BIRBORB99406-20 BIRBORB99406-100
Anti-RAF1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: RAF1 (v-raf-1 murine leukemia viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR