Anti-IKBKG Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB99392-20EA 0 CHF
BIRBORB99392-20 BIRBORB99392-100
Anti-IKBKG Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: IKBKG (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR