Anti-PRKCD Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB99378-100EA 0 CHF
BIRBORB99378-100 BIRBORB99378-20
Anti-PRKCD Rabbit Polyclonal Antibody
Antikörper
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: PRKCD (protein kinase C, delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR