Anti-EFNB1/2 Rabbit polyclonal antibody unconjugated

Supplier: Biorbyt
BIRBORB99366-100EA 0 CHF
BIRBORB99366-100 BIRBORB99366-20
Anti-EFNB1/2 Rabbit polyclonal antibody unconjugated
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: EFNB1/2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR